Learn More
Seracare Life Sciences Inc CONTRAST BLUE SOLUTION
Supplier:  Seracare Life Sciences Inc 55400002
Encompass_Preferred
Contrast BLUE Solution for Immunohistochemistry contains hematoxylin. It is designed for use as a counterstain with the HistoMark immunohistochemical staining systems.
Catalog No. 50-675-56