Learn More
Seracare Life Sciences Inc CONTRAST GREEN SOLUTION
Catalog No. 5067561
Encompass_Preferred
Green nuclear counterstain utilizing methyl green. Contrast Green Solution is available as a component of the HistoMark Black and HistoMark Orange immunohistochemistry substrate kits or as a stand-alone product. Product is also recommended as a counterstain for DAB substrates.
This item is not returnable. View return policy
Catalog No. 50-675-61 Supplier Seracare Life Sciences Inc Supplier No. 55400003
Green nuclear counterstain utilizing methyl green. Contrast Green Solution is available as a component of the HistoMark Black and HistoMark Orange immunohistochemistry substrate kits or as a stand-alone product. Product is also recommended as a counterstain for DAB substrates.