Learn More
Randox Fructosamine Control Level 3, Quality Control reagent, 3 x 1 ml, Human based serum, Lyophilised, Control Level 3, Reconstituted Stability 28 days, Storage 2°C - 8°C
SDP
Catalog No. NC0766327
Encompass_Preferred
The Randox Fructosamine Control Level 3 is a specialized control reagent designed to ensure the accuracy and precision of fructosamine assays in laboratory environments. This lyophilised human serum control offers substantial stability and contributes effectively to compliance with ISO 15189 requirements.
  • Quality control reagent specifically for fructosamine assays
  • Control level 3 for precise performance assessment
  • Lyophilised format enhances stability
  • Human based serum provides reliable results
  • Pack size includes 3 vials of 1 ml each
  • Unreconstituted stability maintained until expiry
  • Unreconstituted storage temperature: 2°C - 8°C
  • Reconstituted stability lasts for 28 days
  • Reconstituted storage temperature: 2°C - 8°C
  • Assayed target values for superior accuracy
  • Aqueous based material ensures product efficacy
  • Reduces operational costs by minimizing waste
Catalog No. NC0766327 Supplier Randox Supplier No. FR2996
The Randox Fructosamine Control Level 3 is a specialized control reagent designed to ensure the accuracy and precision of fructosamine assays in laboratory environments. This lyophilised human serum control offers substantial stability and contributes effectively to compliance with ISO 15189 requirements.
  • Quality control reagent specifically for fructosamine assays
  • Control level 3 for precise performance assessment
  • Lyophilised format enhances stability
  • Human based serum provides reliable results
  • Pack size includes 3 vials of 1 ml each
  • Unreconstituted stability maintained until expiry
  • Unreconstituted storage temperature: 2°C - 8°C
  • Reconstituted stability lasts for 28 days
  • Reconstituted storage temperature: 2°C - 8°C
  • Assayed target values for superior accuracy
  • Aqueous based material ensures product efficacy
  • Reduces operational costs by minimizing waste