Learn More
LifeSensors Control Agarose Beads (negative control)
SDP
Catalog No. 501037770
Encompass_Preferred
Control Agarose Beads. For use in “pre-clearing” cell lysates, as well as a negative control for the pulldown of polyubiquitylated proteins in conjunction with agarose-conjugated TUBEs (UM401 and UM402). Control Agarose is generated by blocking an activated matrix (identical to that used for TUBEs conjugation) with ethanolamine. Control Agarose (200 µl resin) is supplied as a 50% slurry in PBS, pH 7.2, 20% ethanol.
Catalog No. 50-103-7770 Supplier LifeSensors Supplier No. UM04000400
Control Agarose Beads. For use in “pre-clearing” cell lysates, as well as a negative control for the pulldown of polyubiquitylated proteins in conjunction with agarose-conjugated TUBEs (UM401 and UM402). Control Agarose is generated by blocking an activated matrix (identical to that used for TUBEs conjugation) with ethanolamine. Control Agarose (200 µl resin) is supplied as a 50% slurry in PBS, pH 7.2, 20% ethanol.