Learn More
Roche Diagnostics Control B Level One BIO, 1 each
Catalog No. 50589440 Shop All Roche Diagnostics Products
Encompass
Set B consisting of a multi-calibrator and multi-controls for the Cedex Bio and Cedex Bio HT Systems serve for calibration and QC of the tests for Glutamine, and Ammonium
This item is not returnable. View return policy
Catalog No. 50-589-440 Supplier Roche Diagnostics Supplier No. 06682561001
Set B consisting of a multi-calibrator and multi-controls for the Cedex Bio and Cedex Bio HT Systems serve for calibration and QC of the tests for Glutamine, and Ammonium