Learn More
Roche Diagnostics Control B Level Three BIO 1 each
Supplier:  Roche Diagnostics 06682588001
Encompass
Set B consisting of a multi-calibrator and multi-controls for the Cedex Bio and Cedex Bio HT Systems serve for calibration and QC of the tests for Glutamine, and Ammonium
Catalog No. 50-589-442