Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HORIBA Control, BFTROL (Levels 2 and 3)

Catalog No. 22111009 Shop All ABX Diagnostics, Inc. Products
Change view
Click to view available options
Form:
Liquid
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Form
22-111-009 Liquid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-111-009 Supplier HORIBA Supplier No. 1300039405
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Control for hematology analyzer

  • Accessory

Specifications

Sample Type Whole Blood
Type Control
Level 1, 2, 3
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.