Learn More
Med Vet International McKesson TRUE METRIX Glucose Control Solution, 3 mL, 06-R5051-3, Each
Catalog No. 503045351
Encompass_Preferred
Blood Glucose Control Solution McKesson TRUE METRIX 3 mL Level 3 3.0 mL Vial For use with #06-RE4051-43 McKesson TRUE METRIX Self-Monitoring System, #06-R3051-41 and #06-R3051-45 McKesson TRUE METRIX Strips, #06-RE4051P-00 McKesson TRUE METRIX PRO, and #06-R3051P-01 and #06-R3051P-05 McKesson TRUE METRIX PRO Strips
Catalog No. 50-304-5351 Supplier Med Vet International Supplier No. 960303EA
Blood Glucose Control Solution McKesson TRUE METRIX 3 mL Level 3 3.0 mL Vial For use with #06-RE4051-43 McKesson TRUE METRIX Self-Monitoring System, #06-R3051-41 and #06-R3051-45 McKesson TRUE METRIX Strips, #06-RE4051P-00 McKesson TRUE METRIX PRO, and #06-R3051P-01 and #06-R3051P-05 McKesson TRUE METRIX PRO Strips