Learn More
Roche Diagnostics CONTROL C LEVEL 1 BIO
Catalog No. 50589452 Shop All Roche Diagnostics Products
Encompass
Set C consisting of a multi-calibrator and multi-controls for the Cedex Bio and Cedex Bio HT Systems serve for calibration and QC of the tests for Iron, Calcium, Magnesium
Catalog No. 50-589-452 Supplier Roche Diagnostics Supplier No. 07020724001
Set C consisting of a multi-calibrator and multi-controls for the Cedex Bio and Cedex Bio HT Systems serve for calibration and QC of the tests for Iron, Calcium, Magnesium