Learn More
Roche Diagnostics CONTROL C LEVEL 3 BIO
Supplier:  Roche Diagnostics 07020902001
Encompass
Set C consisting of a multi-calibrator and multi-controls for the Cedex Bio and Cedex Bio HT Systems serve for calibration and QC of the tests for Iron, Calcium, Magnesium
Catalog No. 50-589-454