Learn More
MSC HYDAC Polypropylene Vibration Control Clamp, 3/8 in Pipe, 1 ea
Catalog No. NC1910437
Encompass
This HYDAC Polypropylene Standard Duty Vibration Control Clamp is designed for securing 3/8 in pipes. It provides reliable vibration control and includes a carbon steel plate, weld plate, top plate, plastic clamp pair, and bolts for installation.
  • Standard duty clamp type
  • Mounts with weld plate, bolts, clamp pairs, and top plate
  • Constructed from polypropylene clamp material
  • Designed for 3/8 in pipe size
  • Features carbon steel plate material
  • Dimensions of 1.65 in wide x 1.53 in high x 1.18 in deep
Catalog No. NC1910437 Supplier MSC Supplier No. 81921116
This HYDAC Polypropylene Standard Duty Vibration Control Clamp is designed for securing 3/8 in pipes. It provides reliable vibration control and includes a carbon steel plate, weld plate, top plate, plastic clamp pair, and bolts for installation.
  • Standard duty clamp type
  • Mounts with weld plate, bolts, clamp pairs, and top plate
  • Constructed from polypropylene clamp material
  • Designed for 3/8 in pipe size
  • Features carbon steel plate material
  • Dimensions of 1.65 in wide x 1.53 in high x 1.18 in deep