Learn More
Zymo Research Corporation Control E (100 ng/ul) (40 ul)
SDP
Catalog No. 502037169
Encompass_Preferred
Control E DNA (100 ng/µl) (40 µl) for use with Quest 5-hmC DNA ELISA Kit for DNA hydroxymethylation detection and quantification.
Catalog No. 50-203-7169 Supplier Zymo Research Corporation Supplier No. D542555
Control E DNA (100 ng/µl) (40 µl) for use with Quest 5-hmC DNA ELISA Kit for DNA hydroxymethylation detection and quantification.