Learn More
Roche Diagnostics Control E Level 3 Bio, Control reagent for the Cedex Bio and Cedex Bio HT Analyzers, 6 x 0.5mL
Catalog No. NC1648338 Shop All Roche Diagnostics Products
Encompass
Ethanol control solution. This product is intended for quality control by monitoring accuracy and precision of quantitative tests on the Cedex Bio and Cedex Bio HT Analyzers. Control E Level 3 Bio provides an ethanol concentration in the lower part of the measuring range of the ethanol test (ETOHB).
This item is not returnable. View return policy
Catalog No. NC1648338 Supplier Roche Diagnostics Supplier No. 08083827001
Ethanol control solution. This product is intended for quality control by monitoring accuracy and precision of quantitative tests on the Cedex Bio and Cedex Bio HT Analyzers. Control E Level 3 Bio provides an ethanol concentration in the lower part of the measuring range of the ethanol test (ETOHB).