Learn More
Enzo Life Sciences Control : Fc fusion protein (human), (recombinant) (50 µg)
SDP
Supplier:  Enzo Life Sciences ALX203004C050
Encompass_Preferred
Negative control for Fc chimeric proteins. Alternative name: Thy-1 (RLE mutant). MW: ~ 53 kDa. Purity: ≥90% (SDS-PAGE). Formulation: Lyophilized. Contains PBS. Source: Produced in HEK 293 cells. The extracellular domain of mutated hThy-1, hThy-1 (RLE) (aa 20-130), is fused to the Fc portion of human IgG1. The integrin binding sequence RLD of hThy-1 has been changed to RLE abolishing the binding of integrins. Specificity: Does not bind to β3-integrin nor to any of a large number of tested TNF-superfamily members including rhsTRAIL, rhsCD40L, rhsAPRIL, etc. Use/Stability: Stable for at least 6 months after receipt when stored as recommended. Handling: Avoid freeze/thaw cycles. After reconstitution, prepare aliquots and store at -80°C. Long Term Storage: -20°C
Catalog No. NC0892914