Learn More
Aalto Scientific Control FLQ Vitamin D

Description
Control FLQ Vitamin D is an assayed, stable, bi-level reference control for use with assays designed to quantitate Vitamin D. It is intended to simulate human patient samples for the purpose of monitoring the precision of laboratory testing procedures for Vitamin D assays. When used for quality control purposes, it is recommended that each laboratory establish its own means and acceptable ranges and use the values provided only as guides.
Specifications
Specifications
| Detectable Analytes | Vitamin D BP |
| Sample Type | Human Serum |
| Type | Reference Control |
| Disposable | Single-use |
| Form | Frozen Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.