Learn More
Crystal Chem Inc Control for the HDL-Cholesterol Assay Kit, 96-tests
SDP
Catalog No. 501947661
Encompass_Preferred
The Control for the HDL-Cholesterol Assay can be used to validate the calibration curve obtained when running the HDL-Cholesterol Assay. Use this product when: running the assay for the first time, using the kit in a novel way, or when checking the activity of a stored kit.
Catalog No. 50-194-7661 Supplier Crystal Chem Inc Supplier No. 80053
The Control for the HDL-Cholesterol Assay can be used to validate the calibration curve obtained when running the HDL-Cholesterol Assay. Use this product when: running the assay for the first time, using the kit in a novel way, or when checking the activity of a stored kit.