Learn More
Crystal Chem Inc Control for the Mouse HDL-Cholesterol Assay Kit
SDP
Catalog No. 501947660
Encompass_Preferred
The Control for the Mouse HDL Cholesterol Assay can be used to validate the calibration curve obtained when running the Mouse HDL Cholesterol Assay. Use this product when: running the assay for the first time, using the kit in a novel way, or when checking the activity of a stored kit.
Catalog No. 50-194-7660 Supplier Crystal Chem Inc Supplier No. 79993
The Control for the Mouse HDL Cholesterol Assay can be used to validate the calibration curve obtained when running the Mouse HDL Cholesterol Assay. Use this product when: running the assay for the first time, using the kit in a novel way, or when checking the activity of a stored kit.