Learn More
Crystal Chem Inc Control for the Mouse LDL-Cholesterol Assay Kit
SDP
Catalog No. 501947658
Encompass_Preferred
The Control for the Rat LDL Cholesterol Assay can be used to validate the calibration curve obtained when running the Rat LDL Cholesterol Assay. Use this product when: running the assay for the first time, using the kit in a novel way, or when checking the activity of a stored kit.
Catalog No. 50-194-7658 Supplier Crystal Chem Inc Supplier No. 79983
The Control for the Rat LDL Cholesterol Assay can be used to validate the calibration curve obtained when running the Rat LDL Cholesterol Assay. Use this product when: running the assay for the first time, using the kit in a novel way, or when checking the activity of a stored kit.