Learn More
Crystal Chem Inc Control for Rat Creatinine Assay Kit
SDP
Catalog No. 501947691
Encompass_Preferred
The Control for the Rat Creatinine Assay can be used to validate the calibration curve obtained when running the Rat Creatinine Assay. Use this product when: running the assay for the first time, using the kit in a novel way, or when checking the activity of a stored kit.
Catalog No. 50-194-7691 Supplier Crystal Chem Inc Supplier No. 80343
The Control for the Rat Creatinine Assay can be used to validate the calibration curve obtained when running the Rat Creatinine Assay. Use this product when: running the assay for the first time, using the kit in a novel way, or when checking the activity of a stored kit.