Learn More
Crystal Chem Inc Control for Rat Hemoglobin A1c (HbA1c) Assay Kit
SDP
Catalog No. 501947687
Encompass_Preferred
The Control for the Rat Hb1c Assay can be used to validate the calibration curve obtained when running the Rat Hb1c Assay. Use this product when: running the assay for the first time, using the kit in a novel way, or when checking the activity of a stored kit.
Catalog No. 50-194-7687 Supplier Crystal Chem Inc Supplier No. 80303
The Control for the Rat Hb1c Assay can be used to validate the calibration curve obtained when running the Rat Hb1c Assay. Use this product when: running the assay for the first time, using the kit in a novel way, or when checking the activity of a stored kit.