Learn More
Crystal Chem Inc Control for the Total Bile Acids Assay Kit
SDP
Catalog No. 501947681
Encompass_Preferred
The Control for the Total Bile Acid Assay can be used to validate the calibration curve obtained when running the Total Bile Acids Assay. Use this product when: running the assay for the first time, using the kit in a novel way, or when checking the activity of a stored kit.
Catalog No. 50-194-7681 Supplier Crystal Chem Inc Supplier No. 80263
The Control for the Total Bile Acid Assay can be used to validate the calibration curve obtained when running the Total Bile Acids Assay. Use this product when: running the assay for the first time, using the kit in a novel way, or when checking the activity of a stored kit.