Learn More
Grainger Control Key Combination 158 1
Catalog No. 502764613
Encompass_Preferred
The 158KC Control Key by Eaton Cutler-Hammer is a momentary contact key switch. It has a 1/2 inch diameter threaded bushing mount and comes with 2 keys. The electrical rating is 10A 125VAC. This key switch can be used to control access to equipment or machinery. It has an industrial gray finish and is made in the USA. The overall dimensions are 2-1/4 inches x 1-5/8 inches x 1-3/8 inches.
Catalog No. 50-276-4613 Supplier Grainger Supplier No. 16P022
The 158KC Control Key by Eaton Cutler-Hammer is a momentary contact key switch. It has a 1/2 inch diameter threaded bushing mount and comes with 2 keys. The electrical rating is 10A 125VAC. This key switch can be used to control access to equipment or machinery. It has an industrial gray finish and is made in the USA. The overall dimensions are 2-1/4 inches x 1-5/8 inches x 1-3/8 inches.