Learn More
Nanotemper Technologies Inc Monolith NT.115 Control Kit RED
Catalog No. NC1408769
Encompass
Monitor the performance of the Monolith, or use when operating the system for the first time or when training new users. For Nano-GREEN/RED or Nano-BLUE/RED or Pico-RED detectors.
Catalog No. NC1408769 Supplier Nanotemper Technologies Inc Supplier No. MOC030
Monitor the performance of the Monolith, or use when operating the system for the first time or when training new users. For Nano-GREEN/RED or Nano-BLUE/RED or Pico-RED detectors.