Learn More
Med Vet International Diabetes Management Test Control GlucoTrol-AQ Blood Glucose Test Low Level / High Level 2 mL, 180.013.002, Each
Catalog No. 503045532
Encompass_Preferred
GlucoTrol-AQ HemoCue blood glucose test helps in observing the glucose level in the blood of diabetic patients Suitable with HemoCue B-Glucose and HemoCue Glucose 201+ meters and analyzers; yields accurate results; crafted with microbicides to help control growth of micro bacteria Control solution glucose meter comes in two separate vials marked with low level (Level 1) and high level (Level 3) Test solutions are made of microbicides; help control the growth of micro bacteria that otherwise alter glucose concentration Each bottle contains 20 milliliters of solution; for testing low and high glucose levels, respectively
Catalog No. 50-304-5532 Supplier Med Vet International Supplier No. 463324EA
GlucoTrol-AQ HemoCue blood glucose test helps in observing the glucose level in the blood of diabetic patients Suitable with HemoCue B-Glucose and HemoCue Glucose 201+ meters and analyzers; yields accurate results; crafted with microbicides to help control growth of micro bacteria Control solution glucose meter comes in two separate vials marked with low level (Level 1) and high level (Level 3) Test solutions are made of microbicides; help control the growth of micro bacteria that otherwise alter glucose concentration Each bottle contains 20 milliliters of solution; for testing low and high glucose levels, respectively