Learn More
Roche Diagnostics NH3 Bio Test kit for the Cedex Bio Analyzer
Supplier:  Roche Diagnostics 06343775001
Encompass
NH3 Bio Test kit for the Cedex Bio Analyzer. Method: enzymatic amination by glutamate dehydrogenase; recovery 10%
Catalog No. 50-589-379