Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Control liposomes(PBS) | 5 ML
SDP

Supplier:  Medchemexpress LLC HY172202A5ML

Encompass_Preferred

Control Liposomes (PBS) is presented as a white to off-white liquid and is stored at 4°C, without freezing. The product has been tested and complies with the given specifications.

  • Appearance: White to off-white (liquid)
  • Microbiological quality: Conforms
  • Storage: Store at 4°C, do not freeze.

Catalog No. 50-003-85335


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.