Learn More
Crystal Chem Inc Control for the Mouse Creatinine Assay Kit
SDP
Catalog No. 501947695
Encompass_Preferred
The Control for the Mouse Creatinine Assay can be used to validate the calibration curve obtained when running the Mouse Creatinine Assay Assay. Use this product when: running the assay for the first time, using the kit in a novel way, or when checking the activity of a stored kit.
Catalog No. 50-194-7695 Supplier Crystal Chem Inc Supplier No. 80353
The Control for the Mouse Creatinine Assay can be used to validate the calibration curve obtained when running the Mouse Creatinine Assay Assay. Use this product when: running the assay for the first time, using the kit in a novel way, or when checking the activity of a stored kit.