Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Control Normal Human IgG, ProSci
SDP

Catalog No. 16918011 Shop All ProSci Products
Change view
Click to view available options
Quantity:
1.0 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-918-011 1.0 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-918-011 Supplier ProSci LLC Supplier No. 55031000
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Human Control Antibody

Antibody specifically detects in Human, Mouse, Rat Samples. It is validated for ELISA, Western Blot, Immunohistochemistry.

Specifications

Applications ELISA, Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Normal human IgG should be used in immunoassays at the same concentration as the specific human polyclonal antibody.
Formulation PBS containing 0.02% sodium azide.
Host Species Human
Purification Method Normal human IgG is obtained from serum of humans. This preparation can be used as a control reagent for immunoassays. The IgG was purified by DEAE-column chromatography.
Quantity 1.0 mg
Regulatory Status RUO
Target Species Human, Mouse, Rat
Content And Storage Stable at 4°C for three months, store at -20°C for up to one year.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.