Learn More
Roche Diagnostics Control OD Level 2 Bio 6 x 1mL
Catalog No. NC1351996 Shop All Roche Diagnostics Products
Encompass
This product is intended for quality control by monitoring accuracy and precision of the quantitative tests on the Cedex Bio and Cedex Bio HT Analyzers. Control OD Level 2 Bio provides an optical densitiy at 583 nm in the medium part of the measuring range of the OD test.Use this control as described in the instructions of the respective test.
Catalog No. NC1351996 Supplier Roche Diagnostics Supplier No. 07766645001
This product is intended for quality control by monitoring accuracy and precision of the quantitative tests on the Cedex Bio and Cedex Bio HT Analyzers. Control OD Level 2 Bio provides an optical densitiy at 583 nm in the medium part of the measuring range of the OD test.Use this control as described in the instructions of the respective test.