Learn More
Bachem LL-37 (scrambled) Trifluoroacetate
SDP
Supplier:  Bachem 4099707.1000
Encompass_Preferred
LL-37 (scrambled) Trifluoroacetate, H-Gly-Leu-Lys-Leu-Arg-Phe-Glu-Phe-Ser-Lys-Ile-Lys-Gly-Glu-Phe-Leu-Lys-Thr-Pro-Glu-Val-Arg-Phe-Arg-Asp-Ile-Lys-Leu-Lys-Asp-Asn-Arg-Ile-Ser-Val-Gln-Arg-OH, 1354065-56-7, C205H340N60O53, Mr 4493.33, 1mg. Cationic Antimicrobial Protein 18 (134-170) (Scrambled) (human), hCAP-18 (134-170). Control peptide for LL37.
Catalog No. 50-194-7441