Learn More
Siemens Healthineers Control Plasma
Catalog No. 23044683 Shop All Siemens Healthineers Products
Change view
Click to view available options
Type:
Negative Control
Positive Control
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
23-044-683 Negative Control
23-044-695 Positive Control
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-044-683 Supplier Siemens Healthineers Supplier No. 10446235
Add to Cart
Add to Cart
This item is not returnable. View return policy
Assayed control used to monitor the performance of coagulation assays in normal and pathological ranges.
  • Control Plasma P is citrated human plasma from selected healthy blood donors.
    • This control is an assayed control intended to monitor the performance of various analytes in the pathological range.
  • Control Plasma N is citrated normal human pooled plasma from selected healthy blood donors.
    • This control is an assayed control used to monitor the performance of various analytes in the normal range.
TRUSTED_SUSTAINABILITY
Specifications
Type Negative Control
For Use With (Application) Coagulation Testing
For Use With (Equipment) Coagulation Analyzers
Quantity 10 x 1 mL