Learn More
Santa Cruz Biotechnology Control shRNA Lentiviral Particles-A, 200 uL
Catalog No. NC9792178
Encompass
200 ul frozen viral stock containing 1 x 10^6 infectious units of virus (IFU), sufficient for 10-20 transductions. Suitable as a negative control for experiments using targeted shRNA Lentiviral Particles DNA transduction. Encodes of a scrambled shRNA sequence that will not lead to the specific degradation of any cellular message. After transduction, cells stably expressing the control shRNA may be isolated via puromycin selection.
Catalog No. NC9792178 Supplier Santa Cruz Biotechnology Supplier No. SC108080
200 ul frozen viral stock containing 1 x 10^6 infectious units of virus (IFU), sufficient for 10-20 transductions. Suitable as a negative control for experiments using targeted shRNA Lentiviral Particles DNA transduction. Encodes of a scrambled shRNA sequence that will not lead to the specific degradation of any cellular message. After transduction, cells stably expressing the control shRNA may be isolated via puromycin selection.