Learn More
Santa Cruz Biotechnology CONTROL SIRNA
Catalog No. NC9223838
Encompass
Control siRNA (Fluorescein Conjugate)-A, resuspend to 66uL, 10uM, Sufficient for 10 to 20 transfections (in 12 well plate), suitable as a control to monitor transfection efficiency by fluorescence microscopy
Catalog No. NC9223838 Supplier Santa Cruz Biotechnology Supplier No. SC36869
Control siRNA (Fluorescein Conjugate)-A, resuspend to 66uL, 10uM, Sufficient for 10 to 20 transfections (in 12 well plate), suitable as a control to monitor transfection efficiency by fluorescence microscopy