Learn More
Newcomer Supply Copper, Animal Control Slides, 10/set
SDP
Catalog No. NC0313440
Encompass
Newcomer Supply Copper, Animal Control Slides are for the positive histochemical detection of copper in tissue sections. Copper, Animal Control Slides contain sections of positive staining animal liver.
Catalog No. NC0313440 Supplier Newcomer Supply Supplier No. 4130A
Newcomer Supply Copper, Animal Control Slides are for the positive histochemical detection of copper in tissue sections. Copper, Animal Control Slides contain sections of positive staining animal liver.