Learn More
Newcomer Supply Fite, Nocardia sp., Artificial Control Slides, 98/set
SDP
Catalog No. NC0795633
Encompass
Newcomer Supply Fite, Nocardia sp., Artificial Control Slides are for the positive histochemical staining of Nocardia sp. and acid fast bacilli in tissue sections. Nocardia sp., purchased from American Type Culture Collection, is used to produce the positive control tissue. Fite, Nocardia sp., Artificial Control Slides contain sections of positive staining rat lung and negative staining human lung.
Catalog No. NC0795633 Supplier Newcomer Supply Supplier No. 4215B
Newcomer Supply Fite, Nocardia sp., Artificial Control Slides are for the positive histochemical staining of Nocardia sp. and acid fast bacilli in tissue sections. Nocardia sp., purchased from American Type Culture Collection, is used to produce the positive control tissue. Fite, Nocardia sp., Artificial Control Slides contain sections of positive staining rat lung and negative staining human lung.