Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mopec Control Slides, IHC, Box of 100, 98 Unstained, +2 Stained, H. Pylori, Middle Placement
SDP

Catalog No. 22010179 Shop All Mopec Products
Change view
Click to view available options
Control Slide
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
22-010-179 Control Slide
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-010-179 Supplier Mopec Supplier No. SC259100CUST2
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Designed to support the precise demands of laboratories, Mopec Control Slides include Special Stain, IHC, Normal Human Tissue, and Tissue Microarray (TMA).

  • Mopec recognizes the crucial role control slides play in ensuring diagnostic accuracy and reliability, which is why we also provide a unique option for ordering custom control slides, ensuring a precise fit to your specifications.
  • Product specific reactivity is guaranteed for one year from date of receipt when stored properly at 60 to 80°F in a humidity controlled and light-deprived environment.
  • Please revalidate after one year to verify continued reactivity.
  • All control slides are on Mopec IHC Enhanced Hydrophilic Adhesive Slides using Formalin Fixed Paraffin Embedded (FFPE) human tissue.

Specifications

Coating Uncoated
Thickness (Metric) 4 μm
Material Glass
Species Human
Tissue Condition Infected Tissue
Type IHC Control Slide
Dimensions (LxW) 11 x 7 in.
Enhanced Surface No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.