Learn More
Mopec™ Control Slides, Special Stains, Calcium
SDP
Catalog No. 22457686 Shop All Mopec Products
Change view
Click to view available options
Includes:
20 Unstained, 2 Stained Slides
Catalog No. Includes
22-457-686 20 Unstained, 2 Stained Slides
1 options
Catalog No. 22-457-686 Supplier Mopec™ Supplier No. SC80820
Add to Cart
Add to Cart
All positive control slides are on Mopec™ IHC Enhanced Hydrophilic Adhesive Slides using Formalin Fixed Paraffin Embedded (FFPE) human tissue.
  • Product specific reactivity is guaranteed for one year from date of receipt when stored properly at 60°F to 80°F in a humidity controlled and light-deprived environment
  • Case of 10 contains 10 unstained slides and 1 stained slide
  • Case of 20 contains 20 unstained slides and 2 stained slides
  • Case of 100 contains 98 unstained slides and 2 stained slides
Specifications
Includes 20 Unstained, 2 Stained Slides
Product Type Control Slide