Learn More
Associates of Cape Cod Inc CONTROL STANDARD ENDOTOXIN 5PK
SDP
Catalog No. NC9071487
Encompass
CONTROL STANDARD ENDOTOXIN (CSE) FOR PYROCHROME KITS, 10ng/VIAL, 5 VIALS/PACK, reconstitute with LRW ONLY
Catalog No. NC9071487 Supplier Associates of Cape Cod Inc Supplier No. EC0105
CONTROL STANDARD ENDOTOXIN (CSE) FOR PYROCHROME KITS, 10ng/VIAL, 5 VIALS/PACK, reconstitute with LRW ONLY