Learn More
Associates of Cape Cod Inc CONTROL STD ENDOTOXIN, 5/PK
SDP
Catalog No. NC0484725
Encompass
CONTROL STANDARD ENDOTOXIN (CSE), 125ug/VIAL, 5 VIALS/PACK, reconstitute with LRW ONLY
Catalog No. NC0484725 Supplier Associates of Cape Cod Inc Supplier No. E01255
CONTROL STANDARD ENDOTOXIN (CSE), 125ug/VIAL, 5 VIALS/PACK, reconstitute with LRW ONLY