Learn More
Associates of Cape Cod Inc CONTROL STD ENDOTOXIN0.5ML 5PK
SDP
Catalog No. NC0243658
Encompass
CONTROL STANDARD ENDOTOXIN (CSE), 0.5ug/vial, 5 VIALS/PACK, reconstitute with LRW ONLY
Catalog No. NC0243658 Supplier Associates of Cape Cod Inc Supplier No. E00055
CONTROL STANDARD ENDOTOXIN (CSE), 0.5ug/vial, 5 VIALS/PACK, reconstitute with LRW ONLY