Learn More
Charles River Laboratories Control Standard Endotoxin, 10 ng Lipopolysaccharide, Lyophilized powder, 20 EU/mL reconstitution, Storage 2-8°C, Stability 4 weeks after rehydration
Catalog No. NC2824340
Encompass
Control Standard Endotoxin, 10 ng Lipopolysaccharide, Lyophilized powder, 20 EU/mL reconstitution, Storage 2-8°C, Stability 4 weeks after rehydration
Catalog No. NC2824340 Supplier Charles River Laboratories Supplier No. E170
Control Standard Endotoxin, 10 ng Lipopolysaccharide, Lyophilized powder, 20 EU/mL reconstitution, Storage 2-8°C, Stability 4 weeks after rehydration