Learn More
CoaguSense™ Control Strip Kit
GSA_VA
Catalog No. 23654003
Change view
Click to view available options
Quantity:
10
Catalog No. Quantity
23-654-003 10
1 options
Catalog No. 23-654-003 Supplier CoaguSense™ Supplier No. 03P6910
Add to Cart
Add to Cart
CoaguSense™ Control Strip Kit provides extra control strips for facilities requiring more frequent control testing.
  • 5 high control strips
  • 5 low control strips
  • Includes control activation solution
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Content And Storage Room Temperature
Type Liquid Controls
Includes Control activation solution
Quantity 10