Learn More
Med Vet International McKesson Consult Urine Dipstick Control Solution, 2 Levels, 121-011, 6/Box
Catalog No. 503042845
Encompass_Preferred
10 mL For use with McKesson Consult Urine Reagent Strips (121-2GP, 121-4LN, and 121-10SG) For validating visual and analyzer urinalysis readings Level 1 (negative) and Level 2 (positive) Three sets per box (three positive bottles, three negative bottles)
Catalog No. 50-304-2845 Supplier Med Vet International Supplier No. 976927BX
10 mL For use with McKesson Consult Urine Reagent Strips (121-2GP, 121-4LN, and 121-10SG) For validating visual and analyzer urinalysis readings Level 1 (negative) and Level 2 (positive) Three sets per box (three positive bottles, three negative bottles)