Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Controlled Environment Coverall with Boot Cover and Hood

Catalog No. 19165011D Shop All Fisherbrand Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
7X-Large
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-165-011D X-Large
19-165-011A Small
19-165-011B Medium
19-165-011C Large
19-165-011E 2X-Large
19-165-011F 3X-Large
19-165-011G 4X-Large
19-165-011H 5X-Large
19-165-011I 7X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Catalog No. 19-165-011D
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Fisherbrand microporous composite coveralls with hood and boots are designed for many non-hazardous applications and will keep the wearer clean and comfortable all day. Extra coverage and protection with hood and boots.

  • Microporous composite material
  • Strong and lightweight
  • Tapered collar
  • Zipper front
  • Elastic wrist and ankle
  • Elastic hood
  • Anti-skid boots

Specifications

Product Type Cleanroom Coveralls
Size X-Large
Gender Unisex
Material Microporous Composite Fabric
Color White
Disposable Single-use
Waist Style Elastic at Back
Ankle Style Attached Boots
Cuff Style Elastic
Neck Style Hooded
Hood Type Attached, Elastic Face
Closure Type Zipper
Seam Type Serged
Sterility Non-sterile
Resistance Particle
Collar Type Tapered
Latex Free Yes
No. of Pockets None
Sleeve Style Set-in Sleeve
For Use With (Application) Controlled and Critical Environments
Quantity 25/Cs.
Cleanroom Class Class 100 (ISO 5)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.