Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Controlled Environment Frock

Catalog No. 19165010D Shop All Fisherbrand Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
6X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-165-010D X-Large
19-165-010A Small
19-165-010B Medium
19-165-010C Large
19-165-010E 2X-Large
19-165-010F 3X-Large
19-165-010G 4X-Large
19-165-010H 6X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. 19-165-010D
Add to Cart
Add to Cart
This item is not returnable. View return policy

Fisherbrand high quality microporous composite frocks are breathable, strong and lightweight.

  • Microporous composite material
  • Strong, clean and lightweight
  • Mandarin collar
  • Zipper front
  • Elastic wrist
  • Snap collar
  • Zip closure

Specifications

Size X-Large
Material Microporous Composite Fabric
Color White
Disposable Single-use
Sterility Non-sterile
Closure Type Zipper
Cuff Style Elastic
Collar Type Mandarin
Latex Free Yes
No. of Pockets None
Sleeve Style Raglan Sleeve
Packaging Type Double Bagged
Cleanroom Class Class 100 (ISO 5)
Length (English) 46 in.
Product Type Controlled Environments Frock
Seam Type Serged
For Use With (Application) Controlled and Critical Environments
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.