Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Controlled Environment Lab Coat

Catalog No. 19165009E Shop All Fisherbrand Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
5X-Large
7X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-165-009E 2X-Large
19-165-009A Small
19-165-009B Medium
19-165-009C Large
19-165-009D X-Large
19-165-009F 3X-Large
19-165-009G 5X-Large
19-165-009H 7X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. 19-165-009E
Add to Cart
Add to Cart
This item is not returnable. View return policy

Fisherbrand microporous composite lab coat is breathable and lightweight offering comfort and protection for the wearer all day.

  • Microporous composite material
  • Breathable and lightweight
  • Non-confining tapered collar
  • Five snap closure to provide full coverage
  • Elastic wrist
  • Three pockets

Specifications

Product Type Controlled Environments Lab Coat
Size 2X-Large
Gender Unisex
Material Microporous Composite Fabric
Color White
Closure Type Snaps
Cuff Style Elastic
Disposable Single-use
Sterility Non-sterile
Resistance Particulates
No. of Pockets 3
Collar Type Standard/Tapered
Latex Free Yes
Seam Type Serged
Sleeve Style Set-in Sleeve
For Use With (Application) Controlled and Critical Environments
Packaging Type Double Bagged
Autoclavable Not Autoclavable
Cleanroom Class Class 100 (ISO 5)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.