Learn More
Research Products International Corp Controlled Feeding System (Mouse)
SDP
Catalog No. 501977580
Encompass
The Controlled Feeding System is an innovative solution designed to provide precise diet regulation in mouse animal model studies. Its funnel feeder design ensures accurate measurement of feed intake by preventing contamination and feed loss caused by animal handling.
Constructed with a durable polycarbonate enclosure and stainless steel feeding dish, the unit is built for repeated use in demanding laboratory environments. The entire system can be quickly disassembled for thorough cleaning and is fully autoclavable to maintain sterility.
Recommended for mice weighing over 20 grams, the removable tunnel feature also allows accommodation of larger rodents, adding flexibility to study design. Supplied as a complete set, the Controlled Feeding System includes the polycarbonate enclosure and feeder dish, with individual components available separately for replacement or customization.
Catalog No. 50-197-7580 Supplier Research Products International Corp Supplier No. 929102
The Controlled Feeding System is an innovative solution designed to provide precise diet regulation in mouse animal model studies. Its funnel feeder design ensures accurate measurement of feed intake by preventing contamination and feed loss caused by animal handling.
Constructed with a durable polycarbonate enclosure and stainless steel feeding dish, the unit is built for repeated use in demanding laboratory environments. The entire system can be quickly disassembled for thorough cleaning and is fully autoclavable to maintain sterility.
Recommended for mice weighing over 20 grams, the removable tunnel feature also allows accommodation of larger rodents, adding flexibility to study design. Supplied as a complete set, the Controlled Feeding System includes the polycarbonate enclosure and feeder dish, with individual components available separately for replacement or customization.