Learn More
Research Products International Corp Controlled Feeding System, Rat
SDP
Supplier:  Research Products International Corp 929140
Encompass
The Controlled Feeding System is an innovative solution designed to provide precise diet regulation in mouse animal model studies. Its funnel feeder design ensures accurate measurement of feed intake by preventing contamination and feed loss caused by animal handling.
Constructed with a durable polycarbonate enclosure and stainless steel feeding dish, the unit is built for repeated use in demanding laboratory environments. The entire system can be quickly disassembled for thorough cleaning and is fully autoclavable to maintain sterility.
Recommended for rats weighing over 100 grams, the removable tunnel feature also allows accommodation of larger rodents, adding flexibility to study design. Supplied as a complete set, the Controlled Feeding System includes the polycarbonate enclosure and feeder dish, with individual components available separately for replacement or customization.
Catalog No. 50-197-7564