Learn More
Sigma Aldrich Fine Chemicals Biosciences Controlled pore glass - BET specific surface area NIST(R) SRM(R) 2207, nominal pore diameter 18nm | 65997-17-3 |
Catalog No. 501867917
Encompass
Controlled pore glass - BET specific surface area NIST(R) SRM(R) 2207, nominal pore diameter 18nm | 65997-17-3 |
Catalog No. 50-186-7917 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. NIST2207
 Restricted Item
Add to Cart
Add to Cart
Controlled pore glass - BET specific surface area NIST(R) SRM(R) 2207, nominal pore diameter 18nm | 65997-17-3 |