Learn More
Felix Storch Inc ACCUCOLD Controlled Rm Temp. ARG15PV-CRT 15cf GLD
SDP
Catalog No. NC2256203
Encompass_Preferred
ACCUCOLD ARG15PV-CRT (15cf) Controlled Room Temperature (CRT 68F TO 77F) cabinets are designed and purpose-built for ambient medication and vaccine storage in pharmacy and patient care areas. pRE-SET TO 72F. Equipped with Hi/Lo Temp Alarm, Remote Contacts, open door alarm, power failure alarm, Sensor failure alarm, Self closing door, Hospital grade cord & plug, Access Port, Antimicrobial Handle. +/-1.5C Variation The published temperature variation is derived from the maximum deviation of an NTC sensor in a 1 oz. vial located nearest the chamber geometric center during a 24 hour test period. LED interior Light. Glass Door: https://www.accucold.com/product/ARG15PV-CRT
This item is not returnable. View return policy
Catalog No. NC2256203 Supplier Felix Storch Inc Supplier No. ARG15PVCRT
Add to Cart
Add to Cart
Request a Quote
ACCUCOLD ARG15PV-CRT (15cf) Controlled Room Temperature (CRT 68F TO 77F) cabinets are designed and purpose-built for ambient medication and vaccine storage in pharmacy and patient care areas. pRE-SET TO 72F. Equipped with Hi/Lo Temp Alarm, Remote Contacts, open door alarm, power failure alarm, Sensor failure alarm, Self closing door, Hospital grade cord & plug, Access Port, Antimicrobial Handle. +/-1.5C Variation The published temperature variation is derived from the maximum deviation of an NTC sensor in a 1 oz. vial located nearest the chamber geometric center during a 24 hour test period. LED interior Light. Glass Door: https://www.accucold.com/product/ARG15PV-CRT